![Calcifediol for impurity D identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/943/876/08c286a8-8ec3-4ea1-acf9-e0f5668f9edc/640/08c286a8-8ec3-4ea1-acf9-e0f5668f9edc.png)
Calcifediol for impurity D identification
![Calcifediol for impurity D identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/943/876/08c286a8-8ec3-4ea1-acf9-e0f5668f9edc/640/08c286a8-8ec3-4ea1-acf9-e0f5668f9edc.png)
Synonym(s):
25-Hydroxycholecalciferol, 25-Hydroxyvitamin D3
Empirical Formula (Hill Notation):
C27H44O2
CAS Number:
Molecular Weight:
400.64
Products
Product Category
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing