Caffeine-(trimethyl-13C3)
Empirical Formula (Hill Notation):
13C3C5H10N4O2
CAS Number:
Molecular Weight:
197.17
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing