Skip to Content
Merck
Bufexamac European Pharmacopoeia (EP) Reference Standard

Bufexamac

Bufexamac European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2-(p-Butoxyphenyl)acetohydroxamic acid
Empirical Formula (Hill Notation):
C12H17NO3
CAS Number:
Molecular Weight:
223.27

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing