Skip to Content
FR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bone Morphogenetic Protein 2 human
Synonym(s):
BMP-2
Products
Product Category
growth factors (3)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
H4791
Bone Morphogenetic Protein 2 human,
BMP-2, recombinant, expressed in HEK 293 cells, HumanKine, suitable for cell culture
Expand
View Pricing
B3555
Bone Morphogenetic Protein 2 human,
≥98% (SDS-PAGE), recombinant, expressed in
E. coli
, lyophilized powder, suitable for cell culture
Expand
View Pricing
B1814
Bone Morphogenetic Protein 2 human,
Carrier Free, ≥98% (SDS-PAGE), recombinant, expressed in
E. coli
, lyophilized powder, suitable for cell culture
Expand
View Pricing