Skip to Content
Merck
Bis(trifluoromethane)sulfonimide purum, ≥95.0% (19F-NMR)

Bis(trifluoromethane)sulfonimide

Bis(trifluoromethane)sulfonimide purum, ≥95.0% (19F-NMR)
Synonym(s):
Bis(trifluoromethanesulfonyl)amine, Bis(trifluoromethylsulfonyl)amine
Linear Formula:
(CF3SO2)2NH
Molecular Weight:
281.15

Products

Shipping
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing