Skip to Content
Merck
Bisphenol M analytical standard

Bisphenol M

Bisphenol M analytical standard
Synonym(s):
4,4′-(1,3-Phenylenediisopropylidene)bisphenol, Bisphenol M
Linear Formula:
C6H4[C(CH3)2C6H4OH]2
CAS Number:
Molecular Weight:
346.46

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing