![Bisoctrizole Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/251/243/a14d1a4e-8514-457a-860f-c5134a9a1849/640/a14d1a4e-8514-457a-860f-c5134a9a1849.png)
Bisoctrizole Related Compound A
![Bisoctrizole Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/251/243/a14d1a4e-8514-457a-860f-c5134a9a1849/640/a14d1a4e-8514-457a-860f-c5134a9a1849.png)
Synonym(s):
2-(2H-Benzotriazol-2-yl)-4-(1,1,3,3-tetramethylbutyl)phenol, Octrizole
Empirical Formula (Hill Notation):
C20H25N3O
CAS Number:
Molecular Weight:
323.43
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2