![Bis(2,4-dinitrophenyl) oxalate 98%](/deepweb/assets/sigmaaldrich/product/structures/320/581/5d6e4352-9f08-4367-ac58-a39b4dee8975/640/5d6e4352-9f08-4367-ac58-a39b4dee8975.png)
Bis(2,4-dinitrophenyl) oxalate
![Bis(2,4-dinitrophenyl) oxalate 98%](/deepweb/assets/sigmaaldrich/product/structures/320/581/5d6e4352-9f08-4367-ac58-a39b4dee8975/640/5d6e4352-9f08-4367-ac58-a39b4dee8975.png)
Synonym(s):
2,4-Dinitrophenyl oxalate, DNPO, Oxalic acid bis(2,4-dinitrophenyl) ester
Linear Formula:
[-CO2C6H3(NO2)2]2
CAS Number:
Molecular Weight:
422.22
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing