![Betacarotene European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/233/020/e32341ff-4c5b-473b-afd3-0f86ad61c33a/640/e32341ff-4c5b-473b-afd3-0f86ad61c33a.png)
Beta Carotene
![Betacarotene European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/233/020/e32341ff-4c5b-473b-afd3-0f86ad61c33a/640/e32341ff-4c5b-473b-afd3-0f86ad61c33a.png)
Synonym(s):
β-Carotene, β,β-Carotene, Provitamin A
Empirical Formula (Hill Notation):
C40H56
CAS Number:
Molecular Weight:
536.87
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing