Bestatin hydrochloride
Synonym(s):
N-[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine hydrochloride
Empirical Formula (Hill Notation):
C16H24N2O4 · HCl
CAS Number:
Molecular Weight:
344.83
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Activity Spectrum
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing