![Anti-p53 antibody, Mouse monoclonal clone DO-7, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/145/199/ef088b7c-024e-47fb-bba6-22471cac70f1/640/ef088b7c-024e-47fb-bba6-22471cac70f1.jpg)
Anti-p53 antibody, Mouse monoclonal
![Anti-p53 antibody, Mouse monoclonal clone DO-7, purified from hybridoma cell culture](/deepweb/assets/sigmaaldrich/product/images/145/199/ef088b7c-024e-47fb-bba6-22471cac70f1/640/ef088b7c-024e-47fb-bba6-22471cac70f1.jpg)
Synonym(s):
Anti-p53 antibody, Mouse monoclonal, Anti-TRP53, Anti-Transformation-related protein 53
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing