Skip to Content
Merck
Anti-LAMP2 antibody produced in rabbit affinity isolated antibody, buffered aqueous solution

Anti-LAMP2 antibody produced in rabbit

Anti-LAMP2 antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-CD107B, Anti-LGP110, Anti-Lysosomal Membrane Glycoprotein-type B (LGP-B), Anti-Lysosome-Associated Membrane Protein 2, Anti-Cd107b, Anti-Lysosomal-associated membrane protein 2, CD107b, LAMP-2, LAMPB, LGP110

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique