Anti-H6PD antibody produced in rabbit
Synonym(s):
Anti-6PGL)] antibody produced in rabbit, Anti-GDH/6PGL endoplasmic bifunctional protein precursor [Includes: Glucose 1-dehydrogenase antibody produced in rabbit, Anti-Hexose-6-phosphate dehydrogenase); 6-phosphogluconolactonase antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing