
Anti-BRCA2 antibody produced in rabbit

Synonym(s):
Anti-BRCC2, Anti-FACD, Anti-FAD, Anti-FAD1, Anti-FANCD, Anti-FANCD1, Anti-breast cancer 2, early onset, Breast cancer type 2 susceptibility protein
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Post Translational Modification
Species Reactivity
Technique