![Anti-ATR antibody produced in rabbit purified antibody (Ion exchange)](/deepweb/assets/sigmaaldrich/product/images/268/774/92e80393-b5f6-494d-a1fd-05ba9957bf2e/640/92e80393-b5f6-494d-a1fd-05ba9957bf2e.jpg)
Anti-ATR antibody produced in rabbit
![Anti-ATR antibody produced in rabbit purified antibody (Ion exchange)](/deepweb/assets/sigmaaldrich/product/images/268/774/92e80393-b5f6-494d-a1fd-05ba9957bf2e/640/92e80393-b5f6-494d-a1fd-05ba9957bf2e.jpg)
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing