Skip to Content
Merck
Ammonium biborate tetrahydrate 98.9-101.2% B2O3 basis

Ammonium biborate tetrahydrate

Ammonium biborate tetrahydrate 98.9-101.2% B2O3 basis
Synonym(s):
Ammonium tetraborate tetrahydrate
Linear Formula:
(NH4)2B4O7 · 4H2O
CAS Number:
Molecular Weight:
263.38
EC Number:

Products

Brand
Formula Weight
pH Value
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing