
Amlodipine for peak identification

Synonym(s):
Amlodipine besylate, 2-[(2-Aminoethoxy)-methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5- pyridinedicarboxylic acid 3-ethyl 5-methyl ester benzene sulfonate, Norvasc
Empirical Formula (Hill Notation):
C20H25ClN2O5 · C6H5SO3H
CAS Number:
Molecular Weight:
567.05
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing