![Alverine citrate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/198/782/b0cc549a-2c93-447e-92f0-97b0f8a43229/640/b0cc549a-2c93-447e-92f0-97b0f8a43229.png)
Alverine citrate
![Alverine citrate British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/198/782/b0cc549a-2c93-447e-92f0-97b0f8a43229/640/b0cc549a-2c93-447e-92f0-97b0f8a43229.png)
Synonym(s):
Alverine citrate salt, N-Ethyl-3,3′-diphenyldipropylamine, NSC 35459
Empirical Formula (Hill Notation):
C20H27N · C6H8O7
CAS Number:
Molecular Weight:
473.56
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing