Activator 42®
Synonym(s):
5-[3,5-Bis(trifluoromethyl)phenyl]-1H-tetrazole Solution, 5-[3,5-Bis(trifluoromethyl)phenyl]-2H-tetrazole Solution, Activator 42® Solution
CAS Number:
Products
Brand
Color
Physical Form
Product Line
Quality Segment
Showing 1-4 of 4