Skip to Content
Merck
vibrant-m

Acebutolol Related Compound B

vibrant-m
Synonym(s):
N-{3-Acetyl-4-{(2RS)-2-hydroxy-3-[(1-methylethyl)amino]propoxy}phenyl}acetamide, Diacetolol
Empirical Formula (Hill Notation):
C16H24N2O4
CAS Number:
Molecular Weight:
308.37

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1