Skip to Content
Merck
A-769662 ≥98% (HPLC)

A-769662

A-769662 ≥98% (HPLC)
Synonym(s):
4-Hydroxy-3-(2′-hydroxy-1,1′-biphenyl-4-yl)-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carbonitrile, 4-Hydroxy-3-(2′-hydroxybiphenyl-4-yl)-6-oxo-6,7-dihydrothieno[2,3-b]pyridine-5-carbonitrile, 6,7-Dihydro-4-hydroxy-3-(2′-hydroxy[1,1′-biphenyl]-4-yl)-6-oxothieno[2,3-b]pyridine-5-carbonitrile, A 769662, A769662
Empirical Formula (Hill Notation):
C20H12N2O3S
CAS Number:
Molecular Weight:
360.39

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing