Skip to Content
Merck
9-Decenoic acid ≥95%, stabilized, FG

9-Decenoic acid

9-Decenoic acid ≥95%, stabilized, FG
Linear Formula:
H2C=CH(CH2)7CO2H
CAS Number:
Molecular Weight:
170.25
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Chemical Composition
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing