8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt
Synonym(s):
1,3,6-Pyrenetrisulfonic acid, 8-amino trisodium salt, APTS, Trisodium 8-aminopyrene-1,3,6-trisulfonate
Empirical Formula (Hill Notation):
C16H8NNa3O9S3
CAS Number:
Molecular Weight:
523.40
Products
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Showing 1-2 of 2