Skip to Content
Merck
7-[(2-Aminoethyl)amino]-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

7-[(2-Aminoethyl)amino]-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid

7-[(2-Aminoethyl)amino]-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Empirical Formula (Hill Notation):
C15H16FN3O3
CAS Number:
Molecular Weight:
305.30

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade