Skip to Content
Merck
6-Bromo-1-hexanol 97%

6-Bromo-1-hexanol

6-Bromo-1-hexanol 97%
Synonym(s):
Hexamethylene bromohydrin
Linear Formula:
Br(CH2)6OH
CAS Number:
Molecular Weight:
181.07

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing