Skip to Content
Merck
5-Bromo-3-fluoropyridine-2-carboxylic acid 95%

5-Bromo-3-fluoropyridine-2-carboxylic acid

5-Bromo-3-fluoropyridine-2-carboxylic acid 95%
Synonym(s):
5-Bromo-3-fluoropicolinic acid
Empirical Formula (Hill Notation):
C6H3BrFNO2
CAS Number:
Molecular Weight:
220.00

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing