![4-Nitro-o-phenylenediamine analytical standard](/deepweb/assets/sigmaaldrich/product/structures/263/816/cc4218f3-9556-4465-a44d-a3e0e891e1df/640/cc4218f3-9556-4465-a44d-a3e0e891e1df.png)
4-Nitro-o-phenylenediamine
![4-Nitro-o-phenylenediamine analytical standard](/deepweb/assets/sigmaaldrich/product/structures/263/816/cc4218f3-9556-4465-a44d-a3e0e891e1df/640/cc4218f3-9556-4465-a44d-a3e0e891e1df.png)
Synonym(s):
1,2-Diamino-4-nitrobenzene
Linear Formula:
O2NC6H3(NH2)2
CAS Number:
Molecular Weight:
153.14
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Functional Group
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing