4-Methylumbelliferyl sulfate potassium salt
Synonym(s):
Potassium 4-methylumbelliferyl sulfate
Empirical Formula (Hill Notation):
C10H7KO6S
Products
Product Category
Brand
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing