Skip to Content
Merck
4-Isopropylphenol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

4-Isopropylphenol

4-Isopropylphenol certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
p-Cumenol
Linear Formula:
(CH3)2CHC6H4OH
CAS Number:
Molecular Weight:
136.19

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade