
4-Aminophenyl β-D-galactopyranoside

Empirical Formula (Hill Notation):
C12H17NO6
CAS Number:
Molecular Weight:
271.27
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing