
3-O-Acetyl-β-boswellic acid

Synonym(s):
β-Boswellic acid acetate, (3α,4β)-3-(Acetyloxy)urs-12-en-23-oic acid, 3α-Hydroxyurs-12-en-24-oic acid acetate
Empirical Formula (Hill Notation):
C32H50O4
CAS Number:
Molecular Weight:
498.74
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing