Skip to Content
Merck
3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester BioReagent, suitable for fluorescence, ≥97.0% (C)

3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester

3-(4-Hydroxyphenyl)propionic acid N-hydroxysuccinimide ester BioReagent, suitable for fluorescence, ≥97.0% (C)
Synonym(s):
N-Succinimidyl 3-(4-hydroxyphenyl)propionate, N-[(4-Hydroxyhydrocinnamoyl)oxy]succinimide, Bolton-Hunter reagent precursor, Rudinger reagent, SHPP
Empirical Formula (Hill Notation):
C13H13NO5
CAS Number:
Molecular Weight:
263.25
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Product Line
Purity
Application
Markush Class
Markush Group
Reagent Type