Skip to Content
Merck
2-(Trimethylsiloxy)furan 97%

2-(Trimethylsiloxy)furan

2-(Trimethylsiloxy)furan 97%
Empirical Formula (Hill Notation):
C7H12O2Si
CAS Number:
Molecular Weight:
156.25

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing