Skip to Content
Merck
2-(Hydroxymethyl)anthraquinone 97%

2-(Hydroxymethyl)anthraquinone

2-(Hydroxymethyl)anthraquinone 97%
Synonym(s):
2-Hydroxymethyl-9,10-anthracenedione
Empirical Formula (Hill Notation):
C15H10O3
CAS Number:
Molecular Weight:
238.24
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing