Skip to Content
Merck
2-Acetyl-2-thiazoline ≥96%, FG

2-Acetyl-2-thiazoline

2-Acetyl-2-thiazoline ≥96%, FG
Empirical Formula (Hill Notation):
C5H7NOS
CAS Number:
Molecular Weight:
129.18

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing