Skip to Content
Merck
2,6-Dichloropyridine-3-carboxylic acid 97%

2,6-Dichloropyridine-3-carboxylic acid

2,6-Dichloropyridine-3-carboxylic acid 97%
Synonym(s):
2,6-Dichloronicotinic acid
Empirical Formula (Hill Notation):
C6H3Cl2NO2
CAS Number:
Molecular Weight:
192.00

Products

Shipping
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing