Skip to Content
Merck
2,6-Bis(3-(9H-carbazol-9-yl)phenyl)pyridine ≥99% (HPLC)

2,6-Bis(3-(9H-carbazol-9-yl)phenyl)pyridine

2,6-Bis(3-(9H-carbazol-9-yl)phenyl)pyridine ≥99% (HPLC)
Synonym(s):
26DCzPPy
Empirical Formula (Hill Notation):
C41H27N3
CAS Number:
Molecular Weight:
561.67

Products

Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing