1H,1H,2H,2H-Perfluorodecyl acrylate
Synonym(s):
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecyl acrylate, 1H,1H,2H,2H-Perfluorodecyl acrylate
Linear Formula:
H2C=CHCO2CH2CH2(CF2)7CF3
CAS Number:
Molecular Weight:
518.17
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Monomer Type
Reference Material Type
Technique