1-Ethyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide
Synonym(s):
EMIM BTI, EMIM TFSI, EMIMIm
Empirical Formula (Hill Notation):
C8H11F6N3O4S2
CAS Number:
Molecular Weight:
391.31
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-3 of 3