Skip to Content
Merck
1-Benzyl-3-hydroxy-1H-indazole British Pharmacopoeia (BP) Reference Standard

1-Benzyl-3-hydroxy-1H-indazole

1-Benzyl-3-hydroxy-1H-indazole British Pharmacopoeia (BP) Reference Standard
Empirical Formula (Hill Notation):
C14H12N2O
CAS Number:
Molecular Weight:
224.26
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2