![17α-Methyltestosterone solution 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/299/364/58a31360-8f5f-4c06-9bb6-79776292f260/640/58a31360-8f5f-4c06-9bb6-79776292f260.png)
17α-Methyltestosterone solution
![17α-Methyltestosterone solution 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/299/364/58a31360-8f5f-4c06-9bb6-79776292f260/640/58a31360-8f5f-4c06-9bb6-79776292f260.png)
Synonym(s):
17α-Methyl-4-androsten-17β-ol-3-one solution, 17β-Hydroxy-17α-methyl-4-androsten-3-one solution, Mesterone solution, Methyltestosterone solution
Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing