
1-(2-Fluoro-phenyl)-1H-pyrazole-4-carbaldehyde

Empirical Formula (Hill Notation):
C10H7FN2O
Molecular Weight:
190.17
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing