Skip to Content
Merck
1,10-Phenanthroline hydrochloride monohydrate ≥99.5% (titration)

1,10-Phenanthroline hydrochloride monohydrate

1,10-Phenanthroline hydrochloride monohydrate ≥99.5% (titration)
Synonym(s):
o-Phenanthroline hydrochloride monohydrate
Empirical Formula (Hill Notation):
C12H8N2 · HCl · H2O
CAS Number:
Molecular Weight:
234.68
EC Number:

Products

Shipping
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing