Product Name
1-(Cyclopropylmethyl)piperazine 97%
Product Number
658839
Product Brand
aldrich
CAS Number
57184-25-5
Molecular Weight
140.23
Test | Specification |
---|---|
Appearance (Color) | Colorless to Yellow |
Appearance (Form) | Liquid |
Infrared spectrum | Conforms to Structure |
Proton NMR spectra | Conforms to Structure |
Purity (GC) | ≥96.5 % |