Skip to Content
Merck
All Photos(1)

Key Documents

1896

Sigma-Aldrich

Fatty acid standards

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

functional group

carboxylic acid

shipped in

dry ice

storage temp.

-10 to -25°C

General description

Other Notes

100 mg ampule contains approx. equal amounts by weight of:
Tridecanoic acid methyl ester (C13:0)
Pentadecanoic acid methyl ester (C15:0)
Heptadecanoic acid methyl ester (C17:0)
Nonadecanoic acid methyl ester (C19:0)
Heneicosanoic acid methyl ester (C21:0)

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Owen Lonsdale
Zootaxa, 4333(1), 1-85 (2017-12-16)
Family-, genus- and species-level groups in the family Clusiidae (Diptera: Schizophora) are catalogued, providing reference to occurrences of these taxa in the literature, including all those relevant to nomenclature. Full synonymies are provided, including generic combinations for species, and the
The comparative anatomy and pathology of anal glands. Arris and Gale lecture delivered at the Royal College of Surgeons of England on 25th February 1965.
I McColl
Annals of the Royal College of Surgeons of England, 40(1), 36-67 (1967-01-01)
Justin B Runyon
ZooKeys, 966, 57-151 (2020-09-29)
The long-legged flies (Dolichopodidae) of the island of Montserrat in the Lesser Antilles have been surveyed and include 63 species in 27 genera. The following eleven new species are described and illustrated: Amblypsilopus marskeaesp. nov., Medetera ivieisp. nov., Medetera montserratensissp.
Rebeca Martínez et al.
Lipids, 53(4), 429-436 (2018-04-15)
Growth hormone (GH) release is a process that is well regulated by several factors, including GH secretagogues. GH can mediate the regulation of the fatty acid level and composition. The aim of this study was to determine the effect of
Yasushi Kiyokawa et al.
Chemical senses, 29(1), 35-40 (2004-01-31)
Our previous study suggested that the alarm pheromones in rats could be divided into at least two functionally different categories: one evoking autonomic responses and the other evoking behavioral responses, and the present study was conducted to test this hypothesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service