Skip to Content
Merck
All Photos(1)

Key Documents

P9227

Millipore

Potassium tellurite solution

3.5%

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

sterility

sterile-filtered

concentration

3.5%

storage temp.

2-8°C

SMILES string

[K+].[K+].[O-][Te]([O-])=O

InChI

1S/2K.H2O3Te/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

BFPJYWDBBLZXOM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

This product is a concentrated, filtered, sterilized solution [35% (w/v)] of potassium tellurite used to prepare a 3.5% solution, which is recommended for the selective isolation of Staphylococci and Corynebacteria.

Components

Components (1 mL per vial)
Potassium Tellurite 0.35g
Distilled Water 1.0ml

Other Notes

10 mls per vial

Physical form

1 mL of concentrate to prepare 10 mL of 3.5% solution.

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhijie Yang et al.
Journal of bacteriology, 191(11), 3553-3568 (2009-03-31)
The LEE pathogenicity island has been acquired on multiple occasions within the different lineages of enteropathogenic and enterohemorrhagic Escherichia coli. In each lineage, LEE expression is regulated by complex networks of pathways, including core pathways shared by all lineages and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service