Skip to Content
Merck
All Photos(1)

Documents

H9630

Sigma-Aldrich

5-Hydroxymethylcytosine

Synonym(s):

4-Amino-2-hydroxy-5-hydroxymethylpyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N3O2
CAS Number:
Molecular Weight:
141.13
MDL number:
UNSPSC Code:
12352204

SMILES string

Nc1nc(O)ncc1CO

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N W Penn
Archives internationales de pharmacodynamie et de therapie, 218(1), 156-166 (1975-11-01)
The mortality caused by barbiturate poisoning in mice is reduced by the combined action of the DNA pyrimidines thymine and 5-hydroxymethylcytosine (5-HMC) and the structurally related vitamins, pyridoxal, nicotinamide and thiamine, given intraperitoneally. The acute hypnotic effect of sodium pentobarbital

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service