Skip to Content
Merck
All Photos(2)

Documents

MABC1690H

Sigma-Aldrich

Anti-RAINBOW Antibody, HRP conjugated

from mouse, peroxidase conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.45

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

monoclonal

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG1κ

shipped in

ambient

target post-translational modification

unmodified

General description

Pre-stained proteins are commonly used as molecular weight markers in electrophoresis. Several protein markers are available as pre-stained proteins with vinyl sulfone dyes. After separation through electrophoresis, the protein of interest is visualized by specific antibody coupled to a fluorophore or by chemiluminescence detection system. Anti-RAINBOW antibody is a unique antibody that can detect pre-stained molecular markers from different vendors without the need to perform a manual marking and estimating molecular weights. Anti-RAINBOW antibody detects all marker proteins in commercially available RAINBOW markers tested and lacks cross-reactivity with unstained proteins in whole cell lysates of a variety of species. It displays equal detection of Remazol-dye stained proteins: phosphorylase b (red), BSA and ADH (blue), chymotrypsin (yellow) and lysozyme (orange).

Specificity

Anti-RAINBOW, HRP-conjugated is a mouse monoclonal antibody that can be used as a universal tool for visualizing prestained protein molecular weight markers in Western blot analysis. It recognizes and cross-reacts with comparable affinity with four different Remazol dye immunogens.

Immunogen

Purified Remazol Brilliant Blue R (RBBR)-BSA, followed by RO16-Chymotrypsin, RGY-Catalase and RF3B-Lysozyme.

Application

Anti-RAINBOW, HRP-conjugated, Cat. No. MABC1690H, is a mouse monoclonal antibody that recognizes four different Remazol dye-conjugated proteins and can be used to visualize prestained molecular weight markers in Western Blotting.
Research Category
Secondary & Control Antibodies
Western Blotting Analysis: A representative lot detected Remzaol-dye prestained protein standards in Western Blotting applications (Schuchner, S., et. al. (2016). Sci Rep. 6:31363).

Western Blotting Analysis: A 1:500 and 1:1,000 dilution from a representative lot detected protein standard.

Quality

Evaluated by Western Blotting in Blue Pre-Stained Protein Standard.

Western Blotting Analysis: A 1:10,000; 1:20,000; 1:40,000; 1:80,000 dilution of this antibody detected Remzaol-dye prestained protein standards.

Physical form

Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS with StabilZyme® HRP Conjugate Stabilizer and 0.05% Kathon.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

StabilZyme is a registered trademark of SurModics, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lijun Zhao et al.
International journal of molecular medicine, 48(2) (2021-06-17)
The aim of the present study was to identify the function of long non‑coding RNA (lncRNA) small nucleolar RNA host gene 3 (SNHG3) and examine its effects on non‑small cell lung cancer (NSCLC). A series of in vitro experiments were employed to evaluate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service