Skip to Content
Merck
All Photos(1)

Key Documents

6.07233

Supelco

Phenolphthalein

pH indicator

Synonym(s):

Phenolphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14O4
CAS Number:
Molecular Weight:
318.32
MDL number:
UNSPSC Code:
12161500

product name

Phenolphthalein indicator, pH 8.2-9.8

pH

8.2-9.8

InChI

1S/C20H14O4/c21-15-9-5-13(6-10-15)20(14-7-11-16(22)12-8-14)18-4-2-1-3-17(18)19(23)24-20/h1-12,21-22H

InChI key

KJFMBFZCATUALV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Evaluating the effect of diethyl ether and moringa oleifera antioxidant additives on the performance and emission characteristics of jatropha biodiesel-diesel blended fuel on CI engine - An experimental investigation.: This study examines the impact of various additives on biodiesel performance, highlighting the use of phenolphthalein as an indicator in emission analysis (Birhanu et al., 2024).
  • Comparison of detection methods for carbonation depth of concrete.: This research compares various methods for detecting carbonation depth in concrete, including the application of phenolphthalein indicator for assessing chemical changes (Li et al., 2023).
  • Droplet-Impact Driven Formation of Ultralow Volume Liquid Marbles with Enhanced Mechanical Stability and Sensing Ability.: The study explores advanced materials engineering techniques, utilizing phenolphthalein indicator to enhance the sensing capabilities of liquid marbles (Lekshmi et al., 2022).
  • Video colorimetry of single-chromophore systems based on vector analysis in the 3D color space: Unexpected hysteresis loops in oscillating chemical reactions.: This paper presents a novel approach to video colorimetry, employing phenolphthalein as a key chromophore in the analysis (Schell et al., 2020).
  • Webcam camera as a detector for a simple lab-on-chip time based approach.: The study demonstrates the use of a webcam as a detector in microfluidic devices, utilizing phenolphthalein indicator for time-based measurements (Wongwilai et al., 2010).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service