Skip to Content
Merck
All Photos(1)

Key Documents

1.93662

Supelco

Potassium iodate

EMPLURA®

Synonym(s):

Potassium iodate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
IKO3
CAS Number:
Molecular Weight:
214.00
MDL number:
UNSPSC Code:
12352302

product line

EMPLURA®

storage temp.

room temp

SMILES string

[K+].[I](=O)(=O)[O-]

InChI

1S/HIO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

JLKDVMWYMMLWTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service